Skip to Content
Merck
All Photos(7)

Key Documents

T6399

Sigma-Aldrich

Trichloroacetic acid

ACS reagent, ≥99.0%

Synonym(s):

TCA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CCOOH
CAS Number:
Molecular Weight:
163.39
Beilstein:
970119
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

<1 (vs air)

vapor pressure

1 mmHg ( 51 °C)

Assay

≥99.0%

impurities

Substances darkened by sulfuric acid, passes test
≤0.01% insoluble matter

ign. residue

≤0.03%

refractive index

n20/D 1.62 (lit.)

bp

196 °C (lit.)

mp

54-58 °C (lit.)

solubility

H2O: 1 g/10 mL

density

1.62 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.002%
nitrate (NO3-): ≤0.002%
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.002%

functional group

carboxylic acid
chloro

storage temp.

2-8°C

SMILES string

OC(=O)C(Cl)(Cl)Cl

InChI

1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

YNJBWRMUSHSURL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Trichloroacetic acid has been used for the precipitation of sLRP (soluble circulating low density lipoprotein receptor-related protein-1) isolated from plasma.
  • It has been used for the extraction of ascorbic acid from plant samples. It has also been used for the preparation of ascorbate standards for ascorbate assay.
  • It has been used for the precipitation of IMAC (immobilized metal ion affinity chromatography)-isolated phosphoproteins.
Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.

related product

Product No.
Description
Pricing

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Measurement of reduced, oxidized and total ascorbate content in plants.
Gillespie KM and Ainsworth EA
Nature Protocols, 2, 871-871 (2007)
Casein Kinase 2 (CK2)-mediated Phosphorylation of Hsp90? as a Novel Mechanism of Rifampin-induced MDR1 Expression.
Kim SW et al.
The Journal of Biological Chemistry, 290, 17029-17029 (2015)
Impaired lipoprotein receptor-mediated peripheral binding of plasma amyloid-? is an early biomarker for mild cognitive impairment preceding Alzheimer's disease.
Sagare AP et al.
Journal of Alzheimer'S Disease, 24, 25-25 (2011)
Seokjo Kang et al.
Journal of Alzheimer's disease : JAD, 70(3), 667-680 (2019-07-01)
Increased levels of total tau (t-tau) and hyperphosphorylated tau (p-tau) proteins in the cerebrospinal fluid of Alzheimer's disease (AD) patients are well documented and strongly correlate with AD pathology. Recent studies have further shown that human tau can be released
Philip D Campbell et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(44), 14717-14732 (2014-10-31)
Mutations in Kinesin proteins (Kifs) are linked to various neurological diseases, but the specific and redundant functions of the vertebrate Kifs are incompletely understood. For example, Kif5A, but not other Kinesin-1 heavy-chain family members, is implicated in Charcot-Marie-Tooth disease (CMT)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service