Skip to Content
Merck
All Photos(1)

Key Documents

2830-OP

Sigma-Aldrich

Bromophenol Blue Sodium Salt

OmniPur® Grade, Tracking dye for nucleic acid and protein gels

Synonym(s):

OmniPur® Bromophenol Blue, Sodium Salt, Tetrabromophenol Sulfonphthalein, Sodium Salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H9Br4NaO5S
CAS Number:
Molecular Weight:
691.94
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.25
Pricing and availability is not currently available.

Agency

suitable for SM 2310
suitable for SM 2320

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

light red to purple

shipped in

ambient

SMILES string

[Na+].[S]1(=O)(=O)OC(c4c1cccc4)(c3c(cc(cc3Br)[O-])Br)c2c(cc(cc2Br)O)Br

InChI

1S/C19H10Br4O5S.Na/c20-12-5-9(24)6-13(21)17(12)19(18-14(22)7-10(25)8-15(18)23)11-3-1-2-4-16(11)29(26,27)28-19;/h1-8,24-25H;/q;+1/p-1

InChI key

KGANHHDZDYZJEM-UHFFFAOYSA-M

General description

Tracking dye for nucleic acid and protein gels. Suitable for use as a pH indicator or as an lectrophoresis tracking dye for nucleic acids and proteins. Molecular Biology Grade.

Clarity of solution: to pass test
Visual transition (from pH 3.0 to 4.6): Yellow to blue





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Warning

Toxicity: Irritant (B)

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service