Skip to Content
Merck
All Photos(1)

Key Documents

1.05691

Supelco

Lithium hydroxide

98%+, for analysis EMSURE®

Synonym(s):

Lithium hydroxide 98%+

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HLiO
CAS Number:
Molecular Weight:
23.95
MDL number:
UNSPSC Code:
12352305
EC Index Number:
215-183-4
NACRES:
NA.21

form:
solid
solubility:
110 g/L
Pricing and availability is not currently available.

Product Name

Lithium hydroxide 98%+, for analysis EMSURE®

Quality Level

product line

EMSURE®

Assay

≥98.0% (LiOH, acidimetric)

form

solid

potency

210 mg/kg LD50, oral (Rat)

pH

12 (50 °C, 50 g/L in H2O)

mp

423 °C

solubility

110 g/L

density

1.5 g/cm3

Application


  • A study of the hydration of the alkali metal ions in aqueous solution.: This research explores the hydration properties of alkali metal ions, focusing on lithium ions in aqueous solutions. The study provides insights into the hydration structures and dynamics, which are essential for understanding various chemical processes involving lithium hydroxide (Mähler J, Persson I, 2012) (Mähler and Persson, 2012).


  • Determination of markers in digesta and feces by direct current plasma emission spectroscopy.: This paper discusses the use of direct current plasma emission spectroscopy for analyzing markers in biological samples. The method′s applicability for detecting lithium ions among other markers in complex matrices showcases its utility in analytical chemistry and related fields (Combs DK, Satter LD, 1992) (Combs and Satter, 1992).

Analysis Note

Assay (acidimetric, LiOH): ≥ 98.0 %
Carbonate (as Li₂CO₃): ≤ 2.0 %
Chloride (Cl): ≤ 0.05 %
Sulfate (SO₄): ≤ 0.05 %
Heavy metals (as Pb): ≤ 0.002 %
Ca (Calcium): ≤ 0.02 %
Fe (Iron): ≤ 0.002 %
K (Potassium): ≤ 0.02 %
Na (Sodium): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service