Skip to Content
Merck
All Photos(2)

Key Documents

810162P

Avanti

14:0-12:0 NBD PG

Avanti Research - A Croda Brand 810162P, powder

Synonym(s):

1-myristoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-[phospho-rac-(1-glycerol)] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H68N5O13P
CAS Number:
Molecular Weight:
833.95
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (810162P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810162P

shipped in

dry ice

storage temp.

−20°C

General description

NBD PG is a fluorophore having fluorescent 7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) label attached to phosphatidylglycerol (PG). PG is the second most abundant surfactant phospholipid.

Biochem/physiol Actions

7-nitro-2-1,3-benzoxadiazol-4-yl (NBD) labeled fluorescence lipids are used to study the organization of membranes, micelles and reverse micelles. Phosphatidylglycerol (PG) is involved in alveolar stability and plays a vital role in the regulation of innate immune response.

Packaging

5 mL Amber Glass Screw Cap Vial (810162P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marianna Agassandian et al.
Biochimica et biophysica acta, 1831(3), 612-625 (2012-10-03)
Pulmonary surfactant is essential for life and is composed of a complex lipoprotein-like mixture that lines the inner surface of the lung to prevent alveolar collapse at the end of expiration. The molecular composition of surfactant depends on highly integrated
H Raghuraman et al.
Biochimica et biophysica acta, 1768(5), 1258-1267 (2007-03-17)
Lipids that are labeled with the NBD (7-nitrobenz-2-oxa-1,3-diazol-4-yl) group are widely used as fluorescent analogues of native lipids in biological and model membranes to monitor a variety of processes. The NBD group of acyl chain labeled NBD lipids is known

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service