Skip to Content
Merck
All Photos(2)

Key Documents

810156C

Avanti

18:1-12:0 NBD PE

Avanti Research - A Croda Brand 810156C

Synonym(s):

1-oleoyl-2-{12-[(7-nitro-2-1,3-benzoxadiazol-4-yl)amino]dodecanoyl}-sn-glycero-3-phosphoethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H70N5O11P
CAS Number:
Molecular Weight:
840.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 1 mL (810156C-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 810156C

concentration

1 mg/mL (810156C-1mg)

shipped in

dry ice

storage temp.

−20°C

General description

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) is localized at the membrane interface. Fluorescent NBD label is covalently linked to the headgroup of phosphatidylethanolamine. Phosphoethanolamine is mostly present in brain and is the second most abundant, metabolically related aminophospholipid.

Application

18:1-06:0 NBD PE may be used as a component of liposome in cross-linking experiment and to determine M13 coat protein selectivity toward phospholipids headgroups.

Biochem/physiol Actions

N-(7-nitrobenz-2-oxa-1,3-diazol-4-yl) phosphoethanolamine (NBD PE) plays a key role as a fluorescent analog in the studies of organization and dynamics of membranes. NBD PE contributes in monitoring Red edge excitation shift (REES) effects with the unique motional and dielectric properties it possesses.

Packaging

5 mL Amber Glass Screw Cap Vial (810156C-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthetic phosphoethanolamine induces cell cycle arrest and apoptosis in human breast cancer MCF-7 cells through the mitochondrial pathway
Ferreira AK, et al.
Biomedicine / [Publiee Pour L'A.A.I.C.I.G.], 67(6), 481-487 (2013)
The Atg12-Atg5 conjugate has a novel E3-like activity for protein lipidation in autophagy
Hanada T, et al.
Journal of biological and chemical chronicles, 282(52), 37298-37302 (2007)
The Atg12-Atg5 conjugate has a novel E3-like activity for protein lipidation in autophagy
Fernandes F, et al.
Biophysical Journal, 282(52), 37298-37302 (2007)
Cardiolipin and phosphatidylethanolamine role in dibucaine interaction with the mitochondrial membrane
Lopes SC, et al.
Biochimica et Biophysica Acta, 1861(6), 1152-1161 (2019)
Fluorescent methods to study biological membranes, 37-50 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service