Skip to Content
Merck
All Photos(1)

Key Documents

330713X

Avanti

Deuterated Ceramide LIPIDOMIX Mass Spec Standard

Avanti Research - A Croda Brand

Synonym(s):

LIPIDOMIX Mass Spec Standard, Single-vial Prepared Lipidomic Analytical Standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

form

solution (1:1 dichloromethane:methanol)

packaging

ampule of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Packaging

2 mL amber glass sealed ampule

Components

The Deuterated Ceramide LIPIDOMIX Mass Spec Standard is supplied in 1 mL of (1:1) dichloromethane:methanol and contains the following components:

Mixture ComponentTarget Conc. (μM)
  • C16 Ceramide-d7 (d18:1-d7/16:0)40
  • C18 Ceramide-d7 (d18:1-d7/18:0)20
  • C24 Ceramide-d7 (d18:1-d7/24:0)40
  • C24:1 Ceramide-d7 (d18:1-d7/24:1(15Z))20

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
LIPIDOMIX is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weili Pan et al.
Atherosclerosis, 302, 8-14 (2020-05-11)
Ceramides (Cer) are an atherogenic substance. However, the associations between specific plasma Cer levels and culprit plaque morphology in ST-segment elevation myocardial infarction (STEMI) patients are unclear. The study consisted of two parallel cohorts. 100 consecutive patients with STEMI were
Valentina Bosello Travain et al.
Free radical biology & medicine, 147, 80-89 (2019-12-21)
GPx8 is a glutathione peroxidase homolog inserted in the membranes of endoplasmic reticulum (ER), where it seemingly plays a role in controlling redox status by preventing the spill of H2O2. We addressed the impact of GPx8 silencing on the lipidome

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service