Skip to Content
Merck
All Photos(1)

Key Documents

LM6000

Avanti

d5-TG ISTD Mix I

Avanti Research - A Croda Brand LM6000

Synonym(s):

d5-DG ISTD Mix I, d5-TG Internal Standard Mixture I, toluene:methanol (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.25
Pricing and availability is not currently available.

form

ethanol solution

packaging

pkg of 1 × 1 mL (LM6000-1EA)

manufacturer/tradename

Avanti Research - A Croda Brand LM6000

concentration

~10 μg/mL (Refer to C of A for lot specific concentration. )

application(s)

lipidomics
metabolomics

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

d5-TG ISTD Mix I or d5-Triacylglyceride Internal Standard Mixture I has been used as a lipid standard mixture to identify lipids in human serum by tandem mass spectroscopy (MS/MS).[1]

Packaging

2 mL Amber Glass Sealed Ampule (LM6000-1EA)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 1 - STOT SE 3

Target Organs

Central nervous system, Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

44.3 °F

Flash Point(C)

6.85 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ngoc Vu et al.
Analytical and bioanalytical chemistry, 411(23), 5937-5949 (2019-07-08)
Dysregulated lipid species are linked to various disease pathologies and implicated as potential biomarkers for type 1 diabetes (T1D). However, it is challenging to comprehensively profile the blood specimen lipidome with full structural details of every lipid molecule. The commonly
Milka Doktorova et al.
Nature protocols, 13(9), 2086-2101 (2018-09-08)
Freely suspended liposomes are widely used as model membranes for studying lipid-lipid and protein-lipid interactions. Liposomes prepared by conventional methods have chemically identical bilayer leaflets. By contrast, living cells actively maintain different lipid compositions in the two leaflets of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service