Skip to Content
Merck
All Photos(1)

Key Documents

241601C

Avanti

L-α-Phosphatidylcholine

60% (Egg), Avanti Polar Lipids, Chloroform Solution

Synonym(s):

L-α-Phosphatidylcholine (Egg, Chicken-60%) (Total Egg Phosphatide Extract)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Product Name

Egg PC (60%), Avanti Research - A Croda Brand

form

chloroform solution

packaging

pkg of 2 × 20 mL (241601C-1g)

manufacturer/tradename

Avanti Research - A Croda Brand

concentration

25 mg/mL (241601C-1g)

lipid type

phosphoglycerides
phospholipids

shipped in

dry ice

storage temp.

−20°C

General description

Phosphatidylcholine (PC) is the most vital component of the mucosal layer of the colon and a strong bilayer-forming lipid. It is the most common phospholipid in mammalian membranes.
Total Egg Extract is prepared by extracting powdered egg yolk with methanol. The methanol extract is partitioned with chloroform and deionized water, and the chloroform phase containing the organic soluble lipids is concentrated.

Biochem/physiol Actions

Phosphatidylcholine (PC) acts as a surfactant within the mucus to form a hydrophobic surface to prevents bacterial penetrance. It treats fat embolism. Phosphatidylcholine reduces the levels of cholesterol and triglycerides.

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (241601C-1g)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Membranes of Cells (2016)
Integrative Medicine - E-Book (2017)
Efficacy of injections of phosphatidylcholine into fat deposits-a non-surgical alternative to liposuction in body-contouring
Karl GH, et al.
Indian journal of plastic surgery : official publication of the Association of Plastic Surgeons of India, 38(2), 119-119 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service