Skip to Content
Merck
All Photos(1)

Documents

65965

Sigma-Aldrich

3-Methyl-1,3-butanediol

≥97.0% (GC)

Synonym(s):

1,1-Dimethyl-1,3-propanediol, 1,3-Dihydroxy-3-methylbutane, 2-Methyl-2,4-butanediol, 3,3-Dimethyl-1,3-propanediol, 3-Hydroxy-3-methylbutan-1-ol, 3-Hydroxy-3-methylbutanol, 3-Methyl-1,3-butanediol, 3-Methyl-3-hydroxy-1-butyl alcohol, Isoprene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(OH)CH2CH2OH
CAS Number:
Molecular Weight:
104.15
Beilstein:
1209241
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

impurities

≤1% water

refractive index

n20/D 1.442

bp

203-205 °C (lit.)

density

0.976 g/mL at 20 °C (lit.)

SMILES string

CC(C)(O)CCO

InChI

1S/C5H12O2/c1-5(2,7)3-4-6/h6-7H,3-4H2,1-2H3

InChI key

XPFCZYUVICHKDS-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service