Skip to Content
Merck
All Photos(3)

Key Documents

360511

Sigma-Aldrich

4-Methyl-2-pentanone

ACS reagent, ≥98.5%

Synonym(s):

Isobutyl methyl ketone, Isopropylacetone, MIBK, Methyl isobutyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2COCH3
CAS Number:
Molecular Weight:
100.16
Beilstein:
605399
EC Number:
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
Assay:
≥98.5%
bp:
117-118 °C
vapor pressure:
15.7 mmHg ( 20 °C)
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

vapor density

3.5 (vs air)

vapor pressure

15.7 mmHg ( 20 °C)

Assay

≥98.5%

form

liquid

autoignition temp.

840 °F

expl. lim.

1.2-8 %, 93 °F

impurities

≤0.002 meq/g Titr. acid
≤0.100% water

evapn. residue

≤0.0050%

color

APHA: ≤15
clear

refractive index

n20/D 1.395 (lit.)

bp

117-118 °C

mp

−80 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(C)CC(C)=O

InChI

1S/C6H12O/c1-5(2)4-6(3)7/h5H,4H2,1-3H3

InChI key

NTIZESTWPVYFNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Methyl-2-pentanone (Methyl isobutyl ketone (MIBK)) is an organic solvent widely used in various industrial applications. It is used as a solvent for gums, resins, paints, varnishes, lacquers, and nitrocellulose. It is also used as a denaturant and solvent in cosmetic products.

Application

4-Methyl-2-pentanone is used:
  • To prepare (R)-(-)-4-methyl-2-pentanol via asymmetric hydrogenation in the presence of silica-supported starch-polysulfosiloxane-platinum complex (SiO2-ST-Si-S-Pt).
  • In the preparation of jet-fuel-range alkene [C10 and C11 branched alkanes] via aldol condensation reaction with furfural followed by hydrodeoxygenation reaction.
  • Extracting agent for silver, gold and other rare elements from cyanide solutions.

Features and Benefits

4-Methyl-2-pentanone or Methyl isobutyl ketone (MIBK) commonly used diluent or extractant due to its,
  • Low density
  • Viscosity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 2 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

57.2 °F - closed cup

Flash Point(C)

14 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jose Ruiz-Jimenez et al.
Journal of chromatography. A, 1616, 460825-460825 (2020-01-12)
Several calibration approaches were evaluated for the quantitation of volatile organic compounds in air using miniaturized exhaustive and non-exhaustive sampling techniques, such as in-tube extraction (ITEX) and solid phase microextraction (SPME) Arrow. Eleven compounds, 2-ethyl-hexanol, hexanal, nonanal, toluene, ethyl-benzene, methyl
Ginette Truchon et al.
Journal of occupational and environmental hygiene, 3(3), 137-143 (2006-02-09)
Compartmental and physiologically based toxicokinetic modeling coupled with Monte Carlo simulation were used to quantify the impact of biological variability (physiological, biochemical, and anatomic parameters) on the values of a series of bio-indicators of metal and organic industrial chemical exposures.
H J Shim et al.
Journal of chromatography. B, Biomedical applications, 656(2), 407-414 (1994-06-17)
A high-performance liquid chromatographic method was developed for the simultaneous determination of a new anthracycline, DA-125 (I), and its metabolites (M1, M2, M3, and M4) in rat plasma and urine using fluorescein as an internal standard. Compound I, a prodrug
Serife Saçmaci et al.
Analytica chimica acta, 623(1), 46-52 (2008-07-10)
A method for speciation, preconcentration and separation of Fe(II) and Fe(III) in different matrices was developed using solvent extraction and flame atomic absorption spectrometry. 4-Acetyl-5-methyl-1-phenyl-1H-pyrazole-3-carboxylic acid (AMPC) was used as a new complexing reagent for Fe(III). The Fe(III)-AMPC complex was
Sebastian L Riedel et al.
Biotechnology and bioengineering, 110(2), 461-470 (2012-08-21)
Reduced downstream costs, together with high purity recovery of polyhydroxyalkanoate (PHA), will accelerate the commercialization of high quality PHA-based products. In this work, a process was designed for effective recovery of the copolymer poly(hydroxybutyrate-co-hydroxyhexanoate) (P(HB-co-HHx)) containing high levels of HHx

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service