215163
Chloroacetic anhydride
95%
Synonym(s):
Monochloroacetic acid anhydride
About This Item
Recommended Products
vapor pressure
<0.75 mmHg ( 20 °C)
Quality Level
Assay
95%
form
solid
bp
120-123 °C/20 mmHg (lit.)
mp
48-60 °C (lit.)
solubility
chloroform: soluble 100 mg/mL, clear, colorless to faintly yellow
functional group
anhydride
chloro
ester
SMILES string
ClCC(=O)OC(=O)CCl
InChI
1S/C4H4Cl2O3/c5-1-3(7)9-4(8)2-6/h1-2H2
InChI key
PNVPNXKRAUBJGW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Application
- 3,3′-bis(sulfonato)-4,4′-bis(chloroacetamido)azobenzene (BSBCA), a water-soluble, thiol-reactive and photo-switchable cross-linker
- D,L-7-azatryptophan (D,L-7AW)
- 2-methyl-[3,4-di-O-acetyl-6-O-(chloroacetyl)-1,2-dideoxy-α-D-glucopyrano]-[2′,1′:4,5]-2-oxazoline
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
289.4 °F - closed cup
Flash Point(C)
143 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service