Merck
  • Synthesis of 3,3'-bis(sulfonato)-4,4'-bis(chloroacetamido)azobenzene and cysteine cross-linking for photo-control of protein conformation and activity.
Synthesis of 3,3'-bis(sulfonato)-4,4'-bis(chloroacetamido)azobenzene and cysteine cross-linking for photo-control of protein conformation and activity.
Nature protocols (2007-04-05)
Darcy C Burns, Fuzhong Zhang, G Andrew Woolley
ABSTRACT
This protocol describes a procedure for the synthesis of 3,3'-bis(sulfonato)-4,4'-bis(chloroacetamido)azobenzene (BSBCA), a water-soluble, thiol-reactive, photo-switchable cross-linker. In addition, a protocol is outlined for installing the cross-linker in an intramolecular fashion onto proteins bearing two surface-exposed Cys residues. BSBCA is designed to be used as an in vitro activity switch that operates by exerting temporal and reversible photo-control over alpha-helix content within synthetic peptides and recombinant proteins. Synthesis of the cross-linker requires approximately 4.5 d, and cross-linking can be performed in 10-12 h.
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 4.00-4.99 %
Sigma-Aldrich
Sodium hypochlorite solution, purum, ~10% (RT)
Sigma-Aldrich
Sodium hypochlorite solution, reagent grade, available chlorine 10-15 %
Sigma-Aldrich
Acetic anhydride, ACS reagent, ≥98.0%
Sigma-Aldrich
Chloroacetic anhydride, 95%