Z757829
Sartorius filter pipette tips
volume range 50-1200 μL, sterile, pack of 960 ea (10 racks of 96 tips)
Synonym(s):
filter pipette tips, pipetting tips, pipettor tips
About This Item
RNase free
barrier
filter
Recommended Products
material
colorless
colorless tip
polypropylene (Virgin)
sterility
sterile
feature
DNase free
RNase free
barrier
filter
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Sartorius 791211F
maximum volume
1.2 mL
volume range
50-1200 μL
volume
50—1250 μL
binding type
low binding surface
General description
These tips have more space between the sample and the filter than conventional filter tips so you do not need to worry about the sample touching the filter whatever the pipetting technique or type of liquid.
Benefits:
- Pipette foaming liquids like buffers and proteins
- Pipette solvents
- Reverse pipetting
- Multiple dispensing functions of electronic pipettes
Other Notes
Choose from one of the most recent versions:
Certificates of Analysis (COA)