Skip to Content
Merck
All Photos(1)

Key Documents

CLS4809

Corning® Isotip® filtered pipet tips

100-1000 μL, universal fit racked pipet tips, natural, sterile, 100 tips/rack, 1000 tips/case

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.02
volume range:
100-1000 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier: no
disposable
endotoxin free
filter
graduation
packaging: rack
packaging:
rack of 100
manufacturer/tradename:
Corning 4809
Pricing and availability is not currently available.

material

natural tip
polypropylene

sterility

sterile

feature

DNase free
RNase free
barrier: no
disposable
endotoxin free
filter
graduation
packaging: rack

packaging

rack of 100

manufacturer/tradename

Corning 4809

volume range

100-1000 μL

suitability

suitable for (applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling)

compatibility

all popular research-grade pipettors

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Isotip® Filtered Pipet Tips
Corning Isotip filtered pipet tips feature an inert, hydrophobic barrier that prevents aerosolised contaminants from contacting pipettor shafts. All filtered pipet tips are packaged sterile and are certified RNase/DNase-free and nonpyrogenic.

Application

Isotips are ideal for applications where avoidance of cross contamination is critical, such as DNA amplification and radioisotope handling.

Legal Information

Corning is a registered trademark of Corning, Inc.
Isotip is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service