Key Documents
Z743533
Richter Anti-Vibration pads
combi pack, L × W 7.0 in. × 8.5 in. , small, L × W 8.5 in. × 11.0 in. , large, pkg of One of each size
About This Item
UNSPSC Code:
55121611
NACRES:
NB.35
Pricing and availability is not currently available.
manufacturer/tradename
Diversified Biotech VIBE-3000
packaging
pkg of One of each size
L × W
7.0 in. × 8.5 in. , small
8.5 in. × 11.0 in. , large
General description
Inexpensive solution which dampens vibrations generated from lab equipment
Creates a stable surface to keep equipment in place
Fits vortexes, shakers, rockers and microscopes
Small, lightweight design
Stack pads for heavier units or position side-by-side forlarger equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)