Key Documents
Z217050
Sigma-Aldrich
Scienceware® Vibrasorb® vibration damping mount
size 18 in. × 22 in.
Synonym(s):
Vibration dampening mount, Vibration isolator
About This Item
UNSPSC Code:
41100000
NACRES:
NB.71
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H18386-0000
size
18 in. × 22 in.
weight
87 lb
General description
Isolates equipment from disturbing vibrations down to 9 Hz. Molded of dense, 2 ½ in. thick, black and white terrazzo with a polished surface that resists scratches and chemicals. Supported by four Vibro-Absorber cushions and neoprene feet. Overall height is 3 in. Supports 16 kg.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Vibrasorb is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)