Key Documents
Z710970
Rack for glass slides
glass, for 16/32 slides
Synonym(s):
900254
About This Item
UNSPSC Code:
41122800
NACRES:
MA.01
material
glass
feature
Holds 16/32 x slides 76x26 mm
manufacturer/tradename
WHEATON® 900254
size
118x88x70 mm
General description
Glass rack holds 16 single slides, 31 slides arranged alternately straight across and diagonally, or 32 slides back-to-back.
Features and Benefits
  • Manufactured from soda-lime glass
  • These mix-and-match components offer greater flexibility in meeting your requirements
  • This staining dish accommodates 16, 20 and 30 slide racks
  • It holds slides sizes 3” x 1” (75mm x 25mm), 3” x 1-1/2” (75mm x 38mm), and 3” x 2” (75mm x 51mm)
  • Approximate inside bottom dimensions (mm): 121 L x 90 W x 66 D
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)