Key Documents
TTRT96P250BW-EP
Eppendorf twin.tec® Real-Time PCR Plates 96
Semi-Skirted, PCR clean, volume 250 μL, blue plate (white wells), pack of 25
About This Item
UNSPSC Code:
41106300
material:
blue plate (white wells)
polypropylene wells
size:
96 well
sterility:
non-sterile
feature:
skirt
PCR clean
Semi-Skirted
material
blue plate (white wells)
polypropylene wells
sterility
non-sterile
feature
skirt
PCR clean
Semi-Skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951022043 (NA Only)
technique(s)
PCR: suitable
size
96 well
volume
250 μL
working volume
250 μL
Related Categories
General description
The white wells of Eppendorf’s twin.tec real-time PCR plates increase the reflection of fluorescent light in real-time PCR (qPCR) and other fluorescence assays. The increased fluorescence from these specialized qPCR plates improves the sensitivity of your assays, while also improving reproducibility and consistency of your results.
Application
Eppendorf twin.tec®Real-Time PCR Plates 96 is ideal for real time PCR and melting curve analysis.
Features and Benefits
  • White wells ensure better fluorescent light and other fluorescence assays reflection
  • For effective sealing rims are raised
  • Polypropylene wells ensure fast heat transfer and frames are made up of polycarbonate that provides easy and reliable handling
  • Extremely stable mechanically with skirted and semi-skirted frames
  • Reduced wall thickness ensures optimal heat transfer
  • Packed in Eppendorf® forensic DNA grade and lot-certified PCR clean for tested purity
  • With barcode available (upon request)
  • Autoclavable
  • White wells for better reflection
  • High mechanical stability
  • Raised rims for effective sealing
  • Skirted and semi-skirted frames
  • Optimal heat transfer due to reduced wall thickness
  • Autoclavable (121 °C, 20 min)
  • PCR clean and Eppendorf Forensic DNA Grade s available
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)