Skip to Content
Merck
All Photos(1)

Key Documents

EP951020362

Eppendorf twin.tec® PCR Plates 96

semi skirted, size 250 μL, blue, pkg of 25 ea

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NB.22
size:
250 μL
96 well
sterility:
non-sterile
feature:
skirt
semi skirted

sterility

non-sterile

feature

skirt
semi skirted

packaging

pkg of 25 ea

manufacturer/tradename

Eppendorf® 951020362

technique(s)

PCR: suitable

size

250 μL
96 well

color

blue

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Eppendorf twin.tec® PCR plates offer reliability and assurance in every polymerase chain reaction (PCR) assay. These plates guarantee purity, rigidity, color, and various user-friendly features, simplifying your daily workflow and enhancing convenience.
Choose Eppendorf twin.tec PCR plates to achieve reliable PCR results every time. twin.tec plates offer purity, stability, different colors and other simple, intelligent features that make your everyday work easier. The stable polycarbonate frame is well suited for automated and high-throughput processes, and the different colors allow you to quickly organize the individual steps of your workflow.

Application

Eppendorf twin.tec® PCR Plates 96 has been used in:
  • emulsion polymerase chain reaction (PCR)
  • genotyping of single-cell in vitro assay
  • flow cytometry of single cell in vitro assay for genomic DNA extraction
  • droplet digital PCR (ddPCR) for droplet generation
PCR

Features and Benefits

  • Designed with thin-walled polypropylene wells that promote excellent heat transfer to the sample.
  • The plates feature strong polycarbonate frames to ensure durability and rigidity.
  • The raised well rims facilitate superior sealing, effectively minimizing the potential for cross-contamination.
  • All polymerase chain reaction (PCR) consumables are made of high-purity plastics and are ‘PCR clean’. They are batch-tested and certified to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors.
  • Semi-skirted for label and barcode Fits most thermal cyclers 250 µL max. well volume (when used with cap strips)
  • Improved well-to-well tolerance
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Maximum recovery rates for improved assay results
  • Available with barcode (upon request)
  • Ideal for quantitative real-time PCR as well
  • Exceptionally solid and torque-resistant polycarbonate frame
  • OptiTrack® matrix for faster sample identification and fewer pipetting errors
  • Raised well rims for effective sealing, also reduces risk of cross-contamination
  • Batch-tested and certified free of DNA, DNAase, RNase and PCR inhibitors (PCR clean)

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Massanella et al.
Bio-protocol, 5(11) (2015-06-05)
Almost all individuals infected with human immunodeficiency virus (HIV) are also infected with cytomegalovirus (CMV) and Epstein Barr virus (EBV). The aims of our studies have included characterizing and measuring the latent HIV reservoir and understanding the association between asymptomatic
Functional profiling of single CRISPR/Cas9-edited human long-term hematopoietic stem cells
Wagenblast E, et al.
Nature Communications (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service