Skip to Content
Merck
All Photos(2)

Documents

SRP5272

Sigma-Aldrich

SIRT5, active, His tagged human

recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

SIR2L5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17.0-26.5 RLU/min·ng

mol wt

~39 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SIRT5(23408)

General description

SIRT5 is a member of the sirtuin family of proteins which are homologs to the yeast Sir2 protein. Sirtuin family contain a sirtuin core domain and are grouped into four classes with SIRT5 being a member of class III. SIRT5 consists of eight exons and is found in two isoforms which encode a 310 aa and a 299 aa protein, respectively. Human SIRT5 is most predominantly expressed in heart muscle cells and in lymphoblasts. Fluorescence in situ hybridization analysis localized the human SIRT5 gene to chromosome 6p23. SIRT5 can deacetylate cytochrome c, a protein of the mitochondrial intermembrane space with a central function in oxidative metabolism as well as apoptosis initiation.

Biochem/physiol Actions

Sirtuins are NAD+-dependent deacetylases. Sirtuin 5 (SIRT5) catalyzes the degradation of negatively charged acylated substrates. It also possesses desuccinylase, demalonylase and deglutarylase activities. The protein is overexpressed in non-small cell lung cancer (NSCLC) and may have a role in cancer growth.

Physical form

Supplied in 50 mM sodium phosphate, pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1 mM PMSF, 0.2 mM DTT, 25% glycerol.

Preparation Note

After opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assignment of the NAD-dependent deacetylase sirtuin 5 gene (SIRT5) to human chromosome band 6p23 by in situ hybridization.
Mahlknecht U
Cytogenetic and genome research, 112(3-4), 208-212 (2006)
SIRT5 facilitates cancer cell growth and drug resistance in non-small cell lung cancer.
Lu W
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 35(11), 10699-10705 (2014)
Chemical probing of the human sirtuin 5 active site reveals its substrate acyl specificity and peptide-based inhibitors.
Roessler C
Angewandte Chemie (International Edition in English), 53(40), 10728-10732 (2014)
Role of the Substrate Specificity-Defining Residues of Human SIRT5 in Modulating the Structural Stability and Inhibitory Features of the Enzyme.
Yu J
PLoS ONE, 11(3), e0152467-e0152467 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service