Skip to Content
Merck
All Photos(1)

Key Documents

F9182

Sigma-Aldrich

FSL-biotin

Synonym(s):

FSL-CONJ(1Biotin)-SC2-L1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C91H149N17NaO32PS
Molecular Weight:
2079.28
UNSPSC Code:
12352200
NACRES:
NA.32

storage temp.

−20°C

Quality Level

Application

All FSL constructs disperse in biocompatible media and spontaneously and stably incorporate into cell membranes. Cells modified with FSL constructs are known as kodecytes and usually maintain their normal vitality and functionality. FSL-biotin has been specifically designed to create biotinylated live cells (kodecytes) but can also be used to modify other hydrophobic surfaces including fixed cells, virions, and solid phase surfaces. Biotinylated kodecytes can be reacted with a variety of biotin binding protein constructs including fluorescent labeled avidin and avidinylated beads/solid surfaces.

Other Notes

FSL-biotin is a KODE technology construct designed to label hydrophobic surfaces including living cells with biotin. All FSL constructs consist of three essential designable features: a functional component (F), a spacer (S), and a diacyl lipid (L). FSL-biotin is comprised of a monomer of biotin (vitamin B7) representing F, conjugated to a maleimide-bearing carboxymethylglycine based linker (SC2) in turn conjugated to an activated adipate derivative of dioleoylphosphatidylethanolamine (L1).

Legal Information

KODE Constructs are supplied by KODE Biotech Materials Limited under license from KODE Biotech Limited.
KODE is a trademark of KODE Biotech Ltd.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service