Skip to Content
Merck
All Photos(1)

Key Documents

OH30129895

Ohaus® pH Meter

Model ST5000-B, AC/DC input 120 V, US 3-pin plug

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115603
Pricing and availability is not currently available.

material

epoxy shaft

feature

CSA approved

packaging

pack of 1 ea

manufacturer/tradename

Ohaus® 30129895

ORP measuring range

-2000.00-+2000.00 mV

pH measuring range

-2-20 pH unit

temperature measuring range

-30-130 °C

Looking for similar products? Visit Product Comparison Guide

General description

The color touchscreen operates similar to smartphones and tablet computers and provides simple operation of the advanced features, such as one-touch toggling of measurement modes.

Application

pH, oxidation-reduction potential (ORP) measurement

Features and Benefits

3 endpoint modes, continuous measurement mode, up to 9 point calibration, 1,000 measurement memory
Automatic and manual temperature compensation, auto buffer recognition, calibration icon

Legal Information

Ohaus is a registered trademark of Ohaus Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service