The double-junction electrode sensor handles wastewater and solutions containing sulfides, heavy metals, and TRIS. There's no membrane. The sensor can be cleaned with just water.
Recommended Products
manufacturer/tradename
Oakton
measuring range
0-50 °C, accuracy: 1 °C (ATC)
pH measuring range
0.00-14.99 pH unit, accuracy: 0.01 pH unit, resolution: 0.01 pH unit
General description
- Push-button three-point calibration, 4.01, 7.00, 10.00
- Double-junction gel pH electrode
- Powered by four 1.5V alkaline batteries
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
-
Is this probe tris insensitive? Does it require any sort of solution to maintain the membrane?
1 answer-
Helpful?
-