Skip to Content
Merck
All Photos(1)

Documents

N1413

Sigma-Aldrich

NSCI

≥97% (HPLC), solid

Synonym(s):

1-(4-Methoxybenzyl)-5-[2-(pyridin-3-yl-oxymethyl)pyrrolidine-1-sulfonyl]-1H-indole-2,3-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25N3O6S
CAS Number:
Molecular Weight:
507.56
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (HPLC)

form

solid

solubility

DMSO: >10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

COc1ccc(CN2C(=O)C(=O)c3cc(ccc23)S(=O)(=O)N4CCC[C@H]4COc5cccnc5)cc1

InChI

1S/C26H25N3O6S/c1-34-20-8-6-18(7-9-20)16-28-24-11-10-22(14-23(24)25(30)26(28)31)36(32,33)29-13-3-4-19(29)17-35-21-5-2-12-27-15-21/h2,5-12,14-15,19H,3-4,13,16-17H2,1H3/t19-/m0/s1

InChI key

XDHJBIYJRNFDKS-IBGZPJMESA-N

Application

NSCI has been used as an apoptosis inhibitor to study its effects on caspase 9 activity in cultured tick cells. It has also been used as an apoptosis inhibitor in terminal deoxynucleotidyl transferase dUTP nick-end labeling (TUNEL) assay to study anti-CD3-induced apoptosis in the cluster of differentiation 4 (CD4+) and cluster of differentiation 8 (CD8+) T cells from pleural effusions, mononuclear cells (PEMCs).

Biochem/physiol Actions

1-(4-Methoxybenzyl)-5-[2-(pyridin-3-yl-oxymethyl)pyrrolidine-1-sulfonyl]-1H-indole-2,3-dione (NSCI) is a nonpeptide caspase 3 selective inhibitor.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heriberto Prado-Garcia et al.
Cancer immunology, immunotherapy : CII, 61(7), 1065-1080 (2011-12-14)
Lung cancer is the second most common form of cancer and the leading cause of cancer death worldwide. Pleural effusions, containing high numbers of mononuclear and tumor cells, are frequent in patients with advanced stages of lung cancer. We reported
Nieves Ayllón et al.
Infection and immunity, 81(7), 2415-2425 (2013-05-01)
Anaplasma phagocytophilum causes human granulocytic anaplasmosis. Infection with this zoonotic pathogen affects gene expression in both the vertebrate host and the tick vector, Ixodes scapularis. Here, we identified new genes, including spectrin alpha chain or alpha-fodrin (CG8) and voltage-dependent anion-selective
Satu Pekkala et al.
The Biochemical journal, 424(2), 211-220 (2009-09-17)
NAG (N-acetyl-L-glutamate), the essential allosteric activator of the first urea cycle enzyme, CPSI (carbamoyl phosphate synthetase I), is a key regulator of this crucial cycle for ammonia detoxification in animals (including humans). Automated cavity searching and flexible docking have allowed
Dongjin Jeong et al.
eLife, 10 (2021-07-06)
To date, no study has demonstrated that soluble Fas ligand (sFasL)-mediated inflammation is regulated via interaction with Fas in vivo. We found that FasL interacts specifically with tumor necrosis factor receptor superfamily (TNFRSF)10B, also known as death receptor (DR)5. Autoantibody-induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service