Merck
  • Soluble Fas ligand drives autoantibody-induced arthritis by binding to DR5/TRAIL-R2.
Soluble Fas ligand drives autoantibody-induced arthritis by binding to DR5/TRAIL-R2.
eLife (2021-07-06)
Dongjin Jeong, Hye Sung Kim, Hye Young Kim, Min Jueng Kang, Hyeryeon Jung, Yumi Oh, Donghyun Kim, Jaemoon Koh, Sung-Yup Cho, Yoon Kyung Jeon, Eun Bong Lee, Seung Hyo Lee, Eui-Cheol Shin, Ho Min Kim, Eugene C Yi, Doo Hyun Chung
ABSTRACT
To date, no study has demonstrated that soluble Fas ligand (sFasL)-mediated inflammation is regulated via interaction with Fas in vivo. We found that FasL interacts specifically with tumor necrosis factor receptor superfamily (TNFRSF)10B, also known as death receptor (DR)5. Autoantibody-induced arthritis (AIA) was attenuated in FasL (Faslgld/gld)- and soluble FasL (FaslΔs/Δs)-deficient mice, but not in Fas (Faslpr/lpr and Fas-/-)- or membrane FasL (FaslΔm/Δm)-deficient mice, suggesting sFasL promotes inflammation by binding to a Fas-independent receptor. Affinity purification mass spectrometry analysis using human (h) fibroblast-like synovial cells (FLSCs) identified DR5 as one of several proteins that could be the elusive Fas-independent FasL receptor. Subsequent cellular and biochemical analyses revealed that DR5 interacted specifically with recombinant FasL-Fc protein, although the strength of this interaction was approximately 60-fold lower than the affinity between TRAIL and DR5. A microarray assay using joint tissues from mice with arthritis implied that the chemokine CX3CL1 may play an important downstream role of the interaction. The interaction enhanced Cx3cl1 transcription and increased sCX3CL1 production in FLSCs, possibly in an NF-κB-dependent manner. Moreover, the sFasL-DR5 interaction-mediated CX3CL1-CX3CR1 axis initiated and amplified inflammation by enhancing inflammatory cell influx and aggravating inflammation via secondary chemokine production. Blockade of FasL or CX3CR1 attenuated AIA. Therefore, the sFasL-DR5 interaction promotes inflammation and is a potential therapeutic target.
Sigma-Aldrich
SB 203580, solid, ≥98% (HPLC)
Sigma-Aldrich
MG-132, Ready Made Solution, ≥90% (HPLC)
Sigma-Aldrich
BMS-345541, ≥98% (HPLC), powder
Sigma-Aldrich
Anti-Fas Antibody (human, neutralizing), clone ZB4, clone ZB4, Upstate®, from mouse
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
NSCI, ≥97% (HPLC), solid
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder