Skip to Content
Merck
All Photos(2)

Key Documents

I4632

Sigma-Aldrich

2-Iminobiotin

≥98% (TLC)

Synonym(s):

Guanidinobiotin, Hexahydro-2-imino-1H-thieno[3,4-d]imidazole-4-pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17N3O2S
CAS Number:
Molecular Weight:
243.33
Beilstein:
18952
MDL number:
UNSPSC Code:
12352203
PubChem Substance ID:
NACRES:
NA.46

biological source

synthetic (organic)

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

1 M HCl: 50 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

[H][C@]12CS[C@@H](CCCCC(O)=O)[C@@]1([H])NC(=N)N2

InChI

1S/C10H17N3O2S/c11-10-12-6-5-16-7(9(6)13-10)3-1-2-4-8(14)15/h6-7,9H,1-5H2,(H,14,15)(H3,11,12,13)/t6-,7-,9-/m0/s1

InChI key

WWVANQJRLPIHNS-ZKWXMUAHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Iminobiotin is a cyclic guanidino analog of biotin that has high affinity for avidin at high pH (>9) and low affinity at low pH (<6). The proteins that are iminobiotinylated are selectively contained within the avidin column at pH 9 to 11 and may be eluted at pH 4 or by the addition of biotin. This technique may be very useful in the study of cell surface proteins by labeling and selectively recovering the components that are periodate-sensitive from intact human erythrocyte surface. 2-Iminobiotin reversibly inhibits NOS (nitric oxide synthases) via its guanidino group. It inhibits NO biosynthesis and may be important to understand the binding-site interactions for this important class of enzymes. NOS oxidizes the guanidino-nitrogen of L-arginine to produce nitric oxide and L-citrulline.

Application

2-Iminobiotin has been used as a competitive inhibitor to bind to biotic binding sites on streptavidin in order to restore fluorescence signal.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cacha Peeters-Scholte et al.
Experimental brain research, 147(2), 200-208 (2002-11-01)
The purpose of this study was to investigate whether combined inhibition of neuronal and inducible nitric oxide synthase (NOS) by 2-iminobiotin, free radical scavenging by allopurinol, and non-protein-bound iron chelation with deferoxamine improved cerebral oxygenation, electrocortical brain activity, and brain
J Clarkson et al.
Biopolymers, 62(6), 307-314 (2002-02-22)
UV resonance Raman (UVRR) spectroscopy is used to study the binding of biotin and 2-iminobiotin by streptavidin, and the results are compared to those previously obtained from the avidin-biotin complex and new data from the avidin-2-iminobiotin complex. UVRR difference spectroscopy
Cora H A Nijboer et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 27(2), 282-292 (2006-06-01)
We have shown earlier that 2-iminobiotin (2-IB) reduces hypoxia-ischemia (HI)-induced brain damage in neonatal rats, and presumed that inhibition of nitric oxide synthases (NOS) was the underlying mechanism. We now investigated the effect of 2-IB treatment in P7 rat pups
Hiroyuki Inoue et al.
Langmuir : the ACS journal of surfaces and colloids, 21(18), 8354-8359 (2005-08-24)
Layered thin films composed of avidin and 2-iminobiotin-labeled poly(ethyleneimine) (ib-PEI) were prepared by a layer-by-layer deposition of avidin and ib-PEI on a solid surface, and the disintegration induced by changing environmental pH and adding biotin in the solution was studied.
Amesh B Patel et al.
Journal of the American Chemical Society, 126(5), 1318-1319 (2004-02-05)
The strength of a multimolecular system depends on the number of interactions that hold it together. Using dynamic force spectroscopy, we show how the kinetic stability of a system decreases as the number of molecular bonds is increased, as predicted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service