
2-Iminobiotin

Synonym(s):
Guanidinobiotin, Hexahydro-2-imino-1H-thieno[3,4-d]imidazole-4-pentanoic acid
Empirical Formula (Hill Notation):
C10H17N3O2S
CAS Number:
Molecular Weight:
243.33
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing