Skip to Content
Merck
All Photos(1)

Key Documents

GE17-6002-33

HiTrap® IEX Selection Kit

Cytiva 17-6002-33

Synonym(s):

IEX Selection Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56
Pricing and availability is not currently available.

ligand

quaternary amine

description

Ion Exchanger Type (value)

packaging

pkg of 7 ea

manufacturer/tradename

Cytiva 17-6002-33

parameter

42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

6% cross-linked agarose

particle size

45-165 μm

average diameter

90 μm

cleaning

2-14

working range

2-12

suitability

suitable for bioprocess medium

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GE28-4110-07GE29321087GE29-0513-24
parameter

42 psi

parameter

42 psi

parameter

-

parameter

42 psi

matrix

6% cross-linked agarose

matrix

-

matrix

-

matrix

6% cross-linked agarose

packaging

pkg of 7 ea

packaging

pack of 7 × 1 mL

packaging

pkg of 5 × 1 mL

packaging

pack of 1 mL

manufacturer/tradename

Cytiva 17-6002-33

manufacturer/tradename

Cytiva 28-4110-07

manufacturer/tradename

Cytiva 29321087

manufacturer/tradename

Cytiva 29-0513-24

ligand

quaternary amine

ligand

-

ligand

-

ligand

sulphopropyl

General description

HiTrap® IEX Selection Kit consists of seven HiTrap® columns, prepacked with different Sepharose Fast Flow ion exchange chromatography media for screening of the most appropriate media and conditions to use for specific application and development work.

Q, SP, DEAE, and CM Sepharose Fast Flow are based on a robust, 6% highly cross-linked beaded agarose matrix with good flow properties and high loading capacities. ANX Sepharose 4 Fast Flow (high sub) is based on 4% highly cross-linked beaded agarose. This results in a medium with higher porosity, which is particularly useful for the purification of high molecular mass proteins.Q and SP Sepharose XL media have long chains of dextran coupled to a robust, 6% highly cross-linked agarose matrix. The dextran chains increase the exposure of the Q or SP charged groups, which results in very high loading capacities. The active end of the charged group is the same for DEAE Sepharose Fast Flow and ANX Sepharose Fast Flow (high sub), the difference is the length of the carbon chain of the charged group. DEAE Sepharose Fast Flow has a diethylaminoethyl-group bound to the agarose whilst ANX Sepharose 4 Fast Flow (high sub) has a diethylaminopropyl group attached. After choosing the optimal medium, prepacked columns and bulk media are available for larger scale preparative work.

Features and Benefits

  • Fast and easy selection with seven different ion-exchange ligands on Sepharose Fast Flow and Sepharose XL, prepacked in 1 mL HiTrap® columns.
  • Offers a fast, easy and convenient way screen for the optimal media for a given application .
  • Excellent for optimization studies; buffer composition, pH, flow rate, sample loading, and elution scheme can be optimized with small sample quantities prior to scale-up.
  • Designed for use with a syringe, peristaltic pump, or chromatography system such as AKTA design.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol (Q, DEAE, ANX,CM), 20% Ethanol, 0.2 M Sodium Acetate (SP))
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

HiTrap is a registered trademark of Cytiva
Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page provides information about the principles and standard conditions for different purification techniques for the purification of multiprotein complexes with from Cytiva.

This page describes principles and standard conditions for different purification techniques of histidine-tagged proteins using Cytiva products.

This page shows volatile and non-volatile buffer suggestions for anion and cation exchange chromatography.

This page covers the standard ÄKTAdesign configurations for simple IEX chromatography.

See All

Protocols

This page covers the principles and methods of chromatofocusing, a chromatography technique that separates proteins according to differences in their isoelectric point (pI).

This page shows how to perform column packing and preparation for ion exchange chromatography and chromatafocusing when using Tricorn or XK columns available from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service