Skip to Content
Merck
All Photos(1)

Key Documents

94137

Sigma-Aldrich

Hyaluronic acid sodium salt from Streptococcus equi

mol wt 10,000-30,000

Synonym(s):

Sodium hyaluronate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

(Streptococcus equi)

Quality Level

form

powder

mol wt

10,000-30,000

color

white

solubility

water: ~5 g/L

shipped in

wet ice

storage temp.

−20°C

SMILES string

[Na+].N([C@H]1[C@@H](O[C@@H]([C@H]([C@@H]1O[C@@H]4O[C@@H]([C@H]([C@@H]([C@H]4O)O)O)C(=O)O)O)CO)O[C@@H]2[C@H](O[C@H]([C@@H]([C@H]2O)O)O[C@@H]3[C@H]([C@@H](O[C@@H]([C@H]3O)CO)O)NC(=O)C)C(=O)O)C(=O)C

InChI

1S/C28H44N2O23.Na/c1-5(33)29-9-18(11(35)7(3-31)47-25(9)46)49-28-17(41)15(39)20(22(53-28)24(44)45)51-26-10(30-6(2)34)19(12(36)8(4-32)48-26)50-27-16(40)13(37)14(38)21(52-27)23(42)43;/h7-22,25-28,31-32,35-41,46H,3-4H2,1-2H3,(H,29,33)(H,30,34)(H,42,43)(H,44,45);/q;+1/t7-,8-,9-,10-,11-,12-,13+,14+,15-,16-,17-,18-,19-,20+,21+,22+,25-,26+,27-,28-;/m1./s1

InChI key

YWIVKILSMZOHHF-QJZPQSOGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hyaluronic acid, an anionic, nonsulfated glycosaminoglycan (GAG), may be used to study inflammation, cell signaling, cell migration, cell differentiation, tissue engineering and wound-healing.[1] Hyaluronic acid may be used as a substrate to identify and characterize catabolizing enzymes such as hyaluronidase(s).[2] Hyaluronic acid may be used in biomaterials research for the development of biological scaffolds for wound-healing applications and hyaluronic acid fillers in human skin.[3]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuming Zhang et al.
Biomaterials, 35(10), 3243-3251 (2014-01-21)
Hydrogels have been widely used for 3-dimensional (3D) cell culture and tissue regeneration due to their tunable biochemical and physicochemical properties as well as their high water content, which resembles the aqueous microenvironment of the natural extracellular matrix. While many
Cross-linked hyaluronic acid based gel and wound healing.
Richard Koehn et al.
Veterinary surgery : VS, 43(2), 227-227 (2014-02-05)
Naresh Chandra et al.
Viruses, 11(3) (2019-03-16)
Epidemic keratoconjunctivitis (EKC) is a severe ocular disease and can lead to visual impairment. Human adenovirus type-37 (HAdV-D37) is one of the major causative agents of EKC and uses sialic acid (SA)-containing glycans as cellular receptors. Currently, there are no
Naresh Chandra et al.
Viruses, 11(3) (2019-03-16)
Glycans on plasma membranes and in secretions play important roles in infection by many viruses. Species D human adenovirus type 37 (HAdV-D37) is a major cause of epidemic keratoconjunctivitis (EKC) and infects target cells by interacting with sialic acid (SA)-containing
Incorporating facial rejuvenation procedures in the dental practice.
Warren Roberts
Today's FDA : official monthly journal of the Florida Dental Association, 25(2), 50-53 (2013-05-23)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service