NIST2636A
Carbon monoxide in nitrogen
NIST® SRM® 2636a, nominal 250 umol/mol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of cylinder
manufacturer/tradename
NIST®
application(s)
air monitoring
environmental
format
mixture
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
This Standard Reference Material (SRM) is a primary gas mixture for which the amount-of-substance fraction, expressed as concentration, may be related to secondary working standards. This SRM is intended for the calibration of instruments used for carbon monoxide determinations and for other applications. For more information, please refer to the SDS and COA.
SRM 2636A_cert
SRM 2636A_SDS
SRM 2636A_cert
SRM 2636A_SDS
Other Notes
Certified for the analytes listed below.
Carbon monoxide (CO)
Reference/Informational value is provided for the following analytes.
Carbon dioxide (CO2), Water (H2O), Methane (CH4), Total hydrocarbon
See certificate for values and more details at nist.gov/SRM.
Carbon monoxide (CO)
Reference/Informational value is provided for the following analytes.
Carbon dioxide (CO2), Water (H2O), Methane (CH4), Total hydrocarbon
See certificate for values and more details at nist.gov/SRM.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service