Skip to Content
Merck
All Photos(1)

Key Documents

486116

Sigma-Aldrich

Carbon-12C monoxide-16O

13C and 18O depleted, 99.95 atom % 16O, 99.9 atom % 12C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
12C16O
Molecular Weight:
27.99
EC Number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

99.9 atom % 12C
99.95 atom % 16O

form

gas

mass shift

depleted

InChI

1S/CO/c1-2

InChI key

UGFAIRIUMAVXCW-UHFFFAOYSA-N

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

The use of a brass regulator is recommended.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Flam. Gas 1A - Press. Gas Compr. Gas - Repr. 1A - STOT RE 1 Inhalation

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service