Skip to Content
Merck
All Photos(1)

Key Documents

938785

Sigma-Aldrich

Sodium tert-butoxide ChemBeads

Synonym(s):

NaOtBu, Sodium 2-methylpropan-2-olate, Sodium t-butoxide, Sodium tert-butanolate, Sodium tert-butylate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352100
Pricing and availability is not currently available.

form

solid ()

Quality Level

composition

loading of base, 14-16 wt. %

SMILES string

[Na+].CC(C)(C)[O-]

InChI

1S/C4H9O.Na/c1-4(2,3)5;/h1-3H3;/q-1;+1

InChI key

MFRIHAYPQRLWNB-UHFFFAOYSA-N

Application

Sodium tert-butoxide may be used as a safe and effective alternative to sodium hydride for the coupling of hydroxyethyl adenine with diethyl p-toluenesulfonyloxymethanephosphonate to form 9-[2-(diethylphosphonomethoxy)ethyl]adenine (diethyl-PMEA), which is a key intermediate to prepare adefovir dipivoxil.
It can also be used:
- To facilitate the coupling of aryl halides with benzene derivatives in the presence of 10-phenanthroline derivative.
- To activate first-row transition-metal pre-catalysts.
- As a base for the amination of aryl chlorides in the presence of a novel palladacyclic precatalyst.
For general uses, product is also available in powdered form (359270)

Features and Benefits

ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Sol. 1 - Self-heat. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noah P Tu et al.
Angewandte Chemie (International ed. in English), 58(24), 7987-7991 (2019-03-21)
Technologies that enable rapid screening of diverse reaction conditions are of critical importance to methodology development and reaction optimization, especially when molecules of high complexity and scarcity are involved. The lack of a general solid dispensing method for chemical reagents
Ana L Aguirre et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(51), 12981-12986 (2021-07-08)
High-throughput experimentation (HTE) methods are central to modern medicinal chemistry. While many HTE approaches to C-N and Csp2 -Csp2 bonds are available, options for Csp2 -Csp3 bonds are limited. We report here how the adaptation of nickel-catalyzed cross-electrophile coupling of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service