Skip to Content
Merck
All Photos(1)

Key Documents

86501

Supelco

Test mixture 2 for apolar capillary columns according to Grob

suitable for GC, ready-to-use

Synonym(s):

Grob Test Mix for Non-Polar Columns for GC

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

mixture of D(−)-2,3-butanediol, decane, dicyclohexylamine, 2,6-dimethylaniline, 2,6-dimethylphenol, 2-ethylcaproic acid, methyl decanoate, methyl laurate, methyl undecanoate, 1-octanol and dodecane in hexane and methylene chloride

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Mixture for a comprehensive, standardized quality test for apolar glass capillary columns[1][2]

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Chromatography A, 219, 13-13 (1981)
K. Grob, jr. et al.
Journal of Chromatography A, 156, 1-1 (1978)

Questions

  1. What are the components of product 8650?

    1 answer
    1. Product 86501 is composed as follows:
      1.Capric acid methyl ester: 52 mg/L
      2.Undecanoic acid methyl ester: 51 mg/L
      3.Lauric acid methyl ester: 50 mg/L
      4.Decane: 37 mg/L
      5.Dodecane: 38 mg/L
      6.1-Octanol: 46 mg/L
      7.D-(-)-2,3-Butanediol: 82 mg/L
      8.2,6 Dimethylaniline: 42 mg/L
      9. Dicyclohexylamine: 44 mg/L
      10. 2-Ethylhexanoic acid: 52 mg/L
      The solvent is Hexane containing 0.4% Methylene chloride.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service