Skip to Content
Merck
All Photos(2)

Key Documents

43217

Supelco

Decamethylcyclopentasiloxane

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H30O5Si5
CAS Number:
Molecular Weight:
370.77
Beilstein:
1800166
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.396 (lit.)

bp

90 °C/10 mmHg (lit.)

density

0.958 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

C[Si]1(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O1

InChI

1S/C10H30O5Si5/c1-16(2)11-17(3,4)13-19(7,8)15-20(9,10)14-18(5,6)12-16/h1-10H3

InChI key

XMSXQFUHVRWGNA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nilton Dos Santos et al.
Scientific reports, 9(1), 6673-6673 (2019-05-02)
Multiple sclerosis (MS) is an autoimmune and neuroinflammatory disease characterized by demyelination of the Central Nervous System. Immune cells activation and release of pro-inflammatory cytokines play a crucial role in the disease modulation, decisively contributing to the neurodegeneration observed in
Huida Wan et al.
Autophagy, 16(3), 531-547 (2019-06-18)
Mutations in the macroautophagy/autophagy gene WDR45 cause β-propeller protein-associated neurodegeneration (BPAN); however the molecular and cellular mechanism of the disease process is largely unknown. Here we generated constitutive wdr45 knockout (KO) mice that displayed cognitive impairments, abnormal synaptic transmission and
Seon Ju Yeo et al.
Soft matter, 11(8), 1582-1588 (2015-01-17)
Colloidal photonic crystals (CPCs) provide a convenient way to generate structural colour with high stability against degradation under environmental factors. For a number of applications including flexible electronic and energy devices, it is important to generate flexible structural colour that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service