335630
Alcian Blue, pyridine variant
Dye content ≥85 %
Synonym(s):
Alcian Blue-tetrakis(methylpyridinium) chloride
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
form
powder or crystals
Quality Level
composition
Dye content, ≥85%
technique(s)
microbe id | staining: suitable
solubility
water: 1 mg/mL
λmax
616 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[Cl-].[Cl-].[Cl-].[Cl-].C(c1ccc2c3nc(nc4n5[Cu]n6c(nc7nc(nc5c8cc(C[n+]9ccccc9)ccc48)c%10ccc(C[n+]%11ccccc%11)cc7%10)c%12ccc(C[n+]%13ccccc%13)cc%12c6n3)c2c1)[n+]%14ccccc%14
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service