Skip to Content
Merck
All Photos(1)

Key Documents

18597

Supelco

Triethylammonium bicarbonate buffer

1 M, suitable for HPLC, LiChropur

Synonym(s):

Triethylammonium hydrogen carbonate buffer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
3624751
MDL number:
UNSPSC Code:
12161702
NACRES:
NA.21
Pricing and availability is not currently available.

form

liquid

Quality Level

quality

LiChropur

concentration

1 M

technique(s)

HPLC: suitable

impurities

≤0.05% non-volatile matter

pH

8.4-8.6 (20 °C in neat)

density

1.015-1.024

UV absorption

λ: 260 nm Amax: ≤0.04
λ: 270 nm Amax: ≤0.04
λ: 280 nm Amax: ≤0.03
λ: 290 nm Amax: ≤0.02
λ: 500 nm Amax: ≤0.02

storage temp.

2-8°C

SMILES string

OC(O)=O.CCN(CC)CC

InChI

1S/C6H15N.CH2O3/c1-4-7(5-2)6-3;2-1(3)4/h4-6H2,1-3H3;(H2,2,3,4)

InChI key

AFQIYTIJXGTIEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triethylammonium bicarbonate buffer is a commonly used chromatographic eluent, volatile buffer, mobile phase and ion-pairing agent for the purification of nucleic acids and biomolecules.[1]

Application

Buffer for use in ion-exchange chromatography and electrophoresis.
Triethylammonium bicarbonate (TEAB) buffer may be used for studying proteins or noncovalent protein macromolecular complexes under “nondenaturing” conditions by electrospray ionization.[2]

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved preparation of 2 M triethylammonium bicarbonate
Carlson M, et al.
Green Chemistry Letters and Reviews, 8(3-4), 37-39 (2015)
D Lemaire et al.
Analytical chemistry, 73(8), 1699-1706 (2001-05-08)
The use of triethylammonium bicarbonate (TEAB) solution in electrospray mass spectrometry proved to be a very efficient way for studying proteins or noncovalent protein complexes under "nondenaturing" conditions. The low charge states observed in the mass spectra improve the separation
Peng Zhao et al.
Autophagy, 16(12), 2180-2192 (2020-01-28)
In plants, macroautophagy/autophagy has mainly been associated with stress-related processes but how it impacts normal physiological and developmental processes remains largely unexplored. Pollen germination is the critical first step toward fertilization in flowering plants. It is metabolically demanding and relies

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service