Skip to Content
Merck
All Photos(1)

Key Documents

24-0940

Sigma-Aldrich

Pepsin from porcine gastric mucosa

CP

Synonym(s):

Pepsin A, Pepsin from hog stomach

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12000000
grade:
CP

grade

CP

form

powder

mol wt

35 kDa

availability

available only in Japan

UniProt accession no.

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pepsin cleavage can be used to produce F(ab′)2 fragments of antibodies. pepsin at www.sigma-aldrich.com/enzymeexplorer.

Biochem/physiol Actions

Preferential cleavage: hydrophobic and aromatic residues in P1 and P1′ postitions. Cleaves Phe-Val, Gln-His, Glu-Ala, Ala-Leu, Leu-Tyr, Tyr-Leu, Gly-Phe, Phe-Phe and Phe-Tyr bonds in the β chain of insulin

Analysis Note

Optimum pH is 2-4. Active in 4 M urea and 3 M guanidine HCl. Stable at 60 °C. Pepsin is irreversibly inactivated at pH 8.0 - 8.5.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dragana Stanic-Vucinic et al.
Molecular nutrition & food research, 56(12), 1894-1905 (2012-10-16)
The aim of this work was to investigate the effects of high-intensity ultrasound (sonication), on the structure and allergenicity of the major cow's milk allergen, beta-lactoglobulin (BLG). Structural changes upon sonication of BLG were monitored by circular dichroism spectroscopy, tryptophan
Gastric proteinases--structure, function, evolution and mechanism of action.
B Foltmann
Essays in biochemistry, 17, 52-84 (1981-01-01)
Benoît Stijlemans et al.
PLoS pathogens, 7(6), e1002072-e1002072 (2011-06-24)
The African trypanosome Trypanosoma brucei, which persists within the bloodstream of the mammalian host, has evolved potent mechanisms for immune evasion. Specifically, antigenic variation of the variant-specific surface glycoprotein (VSG) and a highly active endocytosis and recycling of the surface
J Tack
Alimentary pharmacology & therapeutics, 22 Suppl 1, 48-54 (2005-07-27)
Gastro-oesophageal reflux disease is defined as the presence of symptoms or lesions that can be attributed to the reflux of gastric contents into the oesophagus. Aspiration and prolonged monitoring studies in humans have shown that reflux of gastric contents is
Chuan-Hsiao Han et al.
Food chemistry, 138(2-3), 923-930 (2013-02-16)
Our previous report showed that yam dioscorin and its peptic hydrolysates exhibit radical scavenging activities; however, the functions of these peptic hydrolases are still under investigation. In this study, the thiol-containing peptides derived from computer-aided simulation of pepsin hydrolysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service