Skip to Content
Merck
All Photos(1)

Key Documents

TX1275

Supelco

Trifluoroacetic acid

Meets Reagent Specifications for testing USP/NF monographs

Synonym(s):

Perfluoroacetic Acid, TFA, Perfluoroacetic Acid, TFA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COOH
CAS Number:
Molecular Weight:
114.02
Beilstein:
742035
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-929-3
Pricing and availability is not currently available.

Agency

meets USP/NF

Quality Level

vapor density

3.9 (vs air)

vapor pressure

97.5 mmHg ( 20 °C)

Assay

≥99.5%

form

liquid

impurities

≤0.3% Water (H2O)

color

APHA: ≤30

refractive index

n20/D 1.3 (lit.)

pH

1 (10 g/L in H2O)

bp

72.4 °C (lit.)

mp

−15.4 °C (lit.)

solubility

soluble 10 g/mL

density

1.489 g/mL at 20 °C (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

OC(C(F)(F)F)=O

InChI

1S/C2HF3O2/c3-2(4,5)1(6)7/h(H,6,7)

InChI key

DTQVDTLACAAQTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Meets Reagent Specifications for testing USP/NF monographs

Analysis Note

Assay: 99.5% min
Color (APHA): 30 max
Form: Clear liquid
Water (H2O): 0.3% max

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

>212.0 °F - Pensky-Martens closed cup

Flash Point(C)

> 100 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service