Skip to Content
Merck
All Photos(2)

Key Documents

HX0078

Supelco

Heptane

greener alternative

Meets Reagent Specifications for testing USP/NF monographs OmniSolv®

Synonym(s):

n-Heptane, high purity solvents

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-563-8
grade:
HPLC grade
technique(s):
HPLC: suitable
application(s):
general analytical
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

grade

HPLC grade

Quality Level

Agency

meets USP/NF

vapor density

3.5 (vs air)

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

product line

OmniSolv®

Assay

≥99% n-heptane basis
≥99.9% (saturated C7 hydrocarbons)

form

liquid

autoignition temp.

433 °F

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤0.01% Water (H2O)
≤5 ng/L Capillary ECD responsive substances (as PCNB)

evapn. residue

≤1 ppm

color

APHA: ≤10

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

transition temp

flash point -4 °C

solubility

0.05 g/L

density

0.684 g/mL at 25 °C (lit.)

UV cutoff

≤197 nm

UV absorption

λ: 197 nm Amax: ≤1.00
λ: 200 nm Amax: ≤0.50
λ: 210 nm Amax: ≤0.25
λ: 220 nm Amax: ≤0.10
λ: 230 nm Amax: ≤0.05
λ: 250 nm Amax: ≤0.01
λ: 260 nm Amax: ≤0.01
λ: 270 nm Amax: ≤0.005
λ: 300 nm Amax: ≤0.005

application(s)

general analytical

greener alternative category

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent compared to hexane and thus align with "Safer Solvents and Auxiliaries". Click here for more information.

Application

HX0078P-1 is packaged in poly coated glass bottles
Meets Reagent Specifications for testing USP/NF monographs OmniSolv®

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (n-heptane)99.0% minAssay (saturated C7 hydrocarbons)99.9% minBoiling range94.5-99.0°CCapillary ECD responsive substances (as PCNB)5ng/L maxColor (APHA)10 maxEvaporation residue1ppm maxFiltered through 0.2 µm filterTo pass testFluorescence (as quinine base)250ppt maxFormClear liquidInfrared SpectrumConforms to standardUV Abs. at 197 nm1.00AU maxUV Abs. at 200 nm0.50AU maxUV Abs. at 210 nm0.25AU maxUV Abs. at 220 nm0.10AU maxUV Abs. at 230 nm0.05AU maxUV Abs. at 250 nm0.01AU maxUV Abs. at 260 nm0.01AU maxUV Abs. at 270 nm0.005AU maxUV Abs. at 300 nm0.005AU maxUV Cut-off197nm maxWater (H2O)0.01% max

Legal Information

OMNISOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service