Skip to Content
Merck
All Photos(1)

Key Documents

SX1310

Supelco

Liquid Acid Spill Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131905
Pricing and availability is not currently available.

pH

11.4 ( in H2O, as aqueous solution)

shipped in

ambient

storage temp.

room temp

Application

SpillSolv

Components

6 x 1kg SpillSolv Acid Neutralizer, 1 pair splash goggles with ventilation, 1 pair nitril gloves A15, 6 H.D. disposal bags 14" x 20" x 4 mil, 4 disposable plastic scoops and scrapers, 1 big sponge for residue clean ups, and 1 treatment guide. Dimensions: Height 13 inches, Length 191/2 inches, Width 7 inches.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service