Skip to Content
Merck
All Photos(1)

Documents

MAB3077

Sigma-Aldrich

Anti-G Protein Giα-2 Antibody, clone L5

clone L5, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L5, monoclonal

species reactivity

bovine, mouse, human, rat, guinea pig

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GNAI2(2771)

Specificity

Gialpha-2. Strong reactivity with bovine (100%), mouse (69%), rat (69%), human (62%) and guinea pig (43%). Specificity confirmed by Western blot versus bovine Galphat, rat Goalpha, rat Gialpha-1, Gialpha-2, rat Gialpha-3, bovine Gsalpha, and murine GoalphaA and GoalphaB.

Immunogen

Recombinant Gialpha-2, purified by the method of Linder (1990).

Application

Anti-G Protein Giα-2 Antibody, clone L5 is an antibody against G Protein Giα-2 for use in WB.
Research Category
Signaling
Research Sub Category
GPCR, cAMP/cGMP & Calcium Signaling
Western blot 1:500 - 1:1000, with detection by 125I-labeled anti-IgG.

Labels a single protein band of 39 - 42 kDa in bovine and rat membranes.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified immunoglobulin, in 0.02M phosphate buffer, 0.25M sodium chloride, pH 7.6, with 0.1% sodium azide.

Storage and Stability

Maintain at 2-8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiromi Ohara et al.
The Journal of veterinary medical science, 75(1), 85-88 (2012-08-11)
In rodents, Gα(i2)-expressing sensory neurons (SNs) that co-express vomeronasal receptor type 1 (V1R) are specifically found in the vomeronasal organ (VNO) and project their axons to the accessory olfactory bulb (AOB). In goats, however, Gα(i2)/V1R-expressing SNs exist in both the
Kati S Mönkkönen et al.
Human reproduction (Oxford, England), 22(5), 1224-1230 (2007-03-10)
Heterotrimeric G proteins take part in membrane-mediated cell signalling and have a role in hormonal regulation. This study clarifies the expression and localization of the G protein subunit G alpha(i2) in the human endometrium and Fallopian tube and changes in
J T O'Flaherty et al.
Journal of immunology (Baltimore, Md. : 1950), 164(6), 3345-3352 (2000-03-08)
5-Oxo-eicosatetraenoic acid (5-oxoETE) stimulated human neutrophil (PMN) and eosinophil chemotaxis, PMN hexose uptake, and PMN membrane GTP/GDP exchange. Pertussis toxin (PT), a blocker of heterotrimeric G proteins (GP), completely inhibited these responses, but proved far less effective on the same
Lucien Gazi et al.
British journal of pharmacology, 138(5), 775-786 (2003-03-19)
(1) The human dopamine D(2long) (D(2L)) receptor was expressed with four different G proteins in Sf9 cells using the baculovirus expression system. When co-expressed with G(i)/G(o) G proteins (G(i1)alpha, G(i2)alpha, G(i3)alpha, or G(o)alpha, plus Gbeta(1) and Ggamma(2)), the receptor displayed
Raghu Ram Katreddi et al.
Development (Cambridge, England), 149(13) (2022-07-06)
The ability of terrestrial vertebrates to find food and mating partners, and to avoid predators, relies on the detection of chemosensory information. Semiochemicals responsible for social and sexual behaviors are detected by chemosensory neurons of the vomeronasal organ (VNO), which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service