Skip to Content
Merck
All Photos(1)

Key Documents

5D41

Millipore

NovaSeptum® GO Conical Tube Sampling Unit with Dip Tube and Inlet C-Flex® Tubing

sterile; γ-irradiated, volume × volume 15 mL × 50 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104019
Pricing and availability is not currently available.

material

TPE tubing (Inlet C-Flex)
polycarbonate Luer lock
polypropylene clip (TTP clip)
polypropylene tube (Fluid contact layer (conical tube))
polystyrene tube (Fluid contact layer (conical tube))
tubing (Female and male Luer-Lok®)

Quality Level

Agency

according to ISO 11137 (sterilization)
according to ISO Clean Room Class 8 conditions in a facility
certified by the ISO 14644-1 standards
meets USP 88 biological reactivity (all component materials)

sterility

sterile; γ-irradiated

product line

NovaSeptum® GO

feature

autoclavable: no

parameter

max. pressure at 0.50 bar (7.25 psi)
2-60 °C temp. range (36-140 °F)

volume × volume

15 mL × 50 mL

application(s)

cell therapy
gene therapy
life science and biopharma
mAb
ophthalmics
parenterals
pharma/biopharma processes
sterile sampling
viral therapy

storage temp.

2-30°C

General description

Device configuration: single Unit Sampling
NovaSeptum® GO Conical Tube Sampling Unit with Dip Tube and Inlet C-Flex® Tubing are small volume sampling containers that fit our established NovaSeptum® GO sampling system. These sampling containers are available in two sizes: 15 mL polystyrene tubes ideal for endotoxin testing, and 50 mL polypropylene tubes for bioburden and other physico-chemical tests. These conical tubes provide flexibility to manufacturers for sampling at any point in the production process eliminating the need to aliquot.

For those samples that need to be transferred between containers, the standard dip tube reaches to the bottom of the tube enabling easy aseptic transfer while minimizing product loss and foaming. When used in a single-use manufacturing process, a weldable option with C-Flex® tubing is available.

The NovaSeptum® GO Conical Tube Sampling Unit offers you flexibility while minimizing the risk of both product and process contamination.

Application

Sterility testing, pyrogenic testing, endotoxin testing, chemical analysis, pH analysis, fermentation applications or any other routine lab testing
supported liquid extraction

Features and Benefits

Fluid contact layer made of polystyrene (15mL), ideal for endotoxin testingSmall size, no need to aliquot, C-Flex inlet weldableDip Tube option:*Reach all the way to the bottom for easy aseptic transfer*Minimizes product loss*Avoids foam creation*Perfect for liquid transfer and bioreactor seeding applications

Packaging

40 units per box

Components

Luer: Polycarbonate

Fluid contact layer (conical tube and dip tube): conical tube: Polystyrene (15mL) and Polypropylene (50mL), dip tube: LDPE (Low density polyethylene pharmalene FL20PH)

Insert Inox 316L

TTP clip Polypropylène (15mL only)

Tubing: Thermoplastic elastomer (TPE), Inlet C-Flex

Legal Information

C-Flex is a registered trademark of Saint-Gobain Performance Plastics Corp.
Luer-Lok is a registered trademark of Becton-Dickinson & Co.
NOVASEPTUM is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

NovaSeptum® GO sterile sampling unit with 15 mL single-use conical tube for small volume and endotoxin sampling. Closed design for aseptic process. Reduce product loss and risks of contamination. Available in single and manifold sampling format. Easily integrated into stainless-steel system via multi-use connection types (TC, in-line, ingold®).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service