Skip to Content
Merck
All Photos(1)

Key Documents

557508

Sigma-Aldrich

Ro-31-7549, Monohydrate

A cell permeable, reversible, selective protein kinase C (PKC) inhibitor (IC₅₀ = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC.

Synonym(s):

Ro-31-7549, Monohydrate, 2-[1-3(Aminopropyl)indol-3-yl]-3(1-methyl-1H-indol-3-yl)maleimide, Acetate, Bisindolylmaleimide VIII, Acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22N4O2 · xC2H4O2 · yH2O
CAS Number:
Molecular Weight:
398.46 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥93% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

red to red-orange

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N1C(=O)C(=C(C1=O)c4c5c([n](c4)C)cccc5)c2c3c([n](c2)CCCN)cccc3

InChI

1S/C24H22N4O2/c1-27-13-17(15-7-2-4-9-19(15)27)21-22(24(30)26-23(21)29)18-14-28(12-6-11-25)20-10-5-3-8-16(18)20/h2-5,7-10,13-14H,6,11-12,25H2,1H3,(H,26,29,30)

InChI key

UQHKJRCFSLMWIA-UHFFFAOYSA-N

General description

A cell permeable, reversible, selective protein kinase C (PKC) inhibitor (IC50 = 158 nM for rat brain PKC) that acts at the ATP binding site of PKC. Does not inhibit the tyrosine phosphorylation or the activation of phospholipase Cγ1. Exhibits some degree of PKC isozyme specificity (IC50 = 53 nM for PKCα, 195 nM for PKCβI, 163 nM for PKCβII, 213 nM for PKCγ, and 175 nM for PKCε). Inhibits carbachol-evoked noradrenaline release (IC50 = 600 nM).
A cell-permeable, reversible, selective protein kinase C (PKC) inhibitor that acts at the ATP binding site of PKC (IC50 = 158 nM for rat brain PKC). Does not inhibit the tyrosine phosphorylation or the activation of phospholipase Cγ1 unlike staurosporine and other serine/threonine kinase inhibitors. IC50 values for individual PKC isozymes are as follows: 53 nM for PKCα, 195 nM for PKCβI, 163 nM for PKCβII, 213 nM for PKCγ, and 175 nM for PKCε. Inhibits carbachol-evoked noradrenaline release (IC50 = 0.6 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
rat brain PKC
Product competes with ATP.
Reversible: yes
Target IC50: 158 nM against rat brain PKC; 53 nM for PKCα, 195 nM for PKCβ, 163 nM for PKCβII, 213 nM for PKCγ, and 175 nM for PKCε

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following initial use, aliquot and refrigerate (4°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Zhang, C., et al. 1997. J. Immunol. 158, 4968.
Turner, N.A., et al. 1996. J. Neurochem. 66, 2381.
Ozawa, K., et al. 1993. J. Biol. Chem. 268, 1749.
Wilkinson, S.E., et al. 1993. Biochem. J. 294, 335.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service