Skip to Content
Merck
All Photos(1)

Key Documents

365251

Sigma-Aldrich

Gö 6983

A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes.

Synonym(s):

Gö 6983, 2-[1-(3-Dimethylaminopropyl)-5-methoxyindol-3-yl]-3-(1H-indol-3-yl) maleimide, Go 6983

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H26N4O3
CAS Number:
Molecular Weight:
442.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange-red

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N1C(=O)C(=C(C1=O)c4c5c([nH]c4)cccc5)c2c3c([n](c2)CCCN(C)C)ccc(c3)OC

InChI

1S/C26H26N4O3/c1-29(2)11-6-12-30-15-20(18-13-16(33-3)9-10-22(18)30)24-23(25(31)28-26(24)32)19-14-27-21-8-5-4-7-17(19)21/h4-5,7-10,13-15,27H,6,11-12H2,1-3H3,(H,28,31,32)

InChI key

LLJJDLHGZUOMQP-UHFFFAOYSA-N

General description

A potent, cell-permeable, inhibitor of protein kinase C (PKC) that has been shown to selectively inhibit several PKC isozymes (IC50 = 7 nM for PKCα and PKCβ; 6 nM for PKCγ; 10 nM for PKCδ; 60 nM for PKCζ). Gö 6983 does not effectively inhibit PKCµ (IC50 = 20 µM) and can thus be used to differentiate PKCµ from other PKC isozymes.
A potent, cell-permeable, reversible, and ATP-competitive inhibitor of protein kinase C (PKC) that inhibits several PKC isozymes (IC50 = 7 nM for PKCα and PKCβ; 6 nM for PKCγ; 10 nM for PKCδ; and 60 nM for PKCζ). Gö 6983 does not effectively inhibit PKCµ (IC50 = 20 µM) and can thus be used to differentiate PKCµ from other PKC isozymes.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCα and PKCβ
Product competes with ATP.
Reversible: yes
Target IC50: 7 nM for PKCα and PKCβ; 6 nM for PKCγ; 10 nM for PKCδ; and 60 nM for PKCζ

Warning

Toxicity: Standard Handling (A)

Other Notes

Wang, D., et al. 1998. J. Biol. Chem. 273, 33027.
Stempka, L., et al. 1997. J. Biol. Chem. 272, 6805.
Gschwendt, M., et al. 1996. FEBS Lett. 392, 77.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Justin Brumbaugh et al.
Nature protocols, 1(2), 1044-1055 (2007-04-05)
Here we describe protocols for preparing and using fluorescent probes that respond to conformational changes by altered Foerster resonance energy transfer (FRET) efficiencies upon phosphorylation or, in principle, other posttranslational modifications (PTMs). The sensor protein, a truncated version of pleckstrin
Ping Jiang et al.
Clinical and experimental pharmacology & physiology, 44(7), 760-770 (2017-04-11)
Tiron functions as an effective antioxidant alleviating the intracellular reactive oxygen species (ROS) or the acute toxic metal overload. Previous studies have shown that cardiac myocyte apoptosis can be effectively inhibited by tiron administration in streptozotocin (STZ)-induced diabetic rats, primary
Alice Liu et al.
PloS one, 15(7), e0236175-e0236175 (2020-07-23)
Adenoviruses cause upper respiratory infections, conjunctivitis, keratitis, and gastrointestinal illness. These can be fatal in immunocompromised individuals. Adenoviruses have also been engineered into viral vectors to deliver therapeutic genes or induce immunity as vaccine carriers. The success of ocular gene
Dilip Kumar et al.
Journal of immunology (Baltimore, Md. : 1950), 182(2), 1011-1020 (2009-01-07)
The MAPKs ERK, JNK, and p38 control diverse aspects of the immune response, including regulation of cytotoxin biology in NK cells and CTL. The chemokine CCL5 is coreleased with the cytotoxins, perforin, the granzymes, and granulysin, during the lethal hit
Vicky García-Hernández et al.
Journal of cellular physiology, 230(1), 105-115 (2014-06-10)
Epidermal Growth Factor (EGF) is a key regulator of epithelial paracellular permeability, a property that depends on tight junctions (TJ) and can be evaluated through the measurement of the transepithelial electrical resistance (TER). EGF increases the TER of MDCK monolayers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service