Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.99006
Buffer solutions
traceable to NIST, traceable to PTB, pH 4.01 (25 °C, phthalate), pH 7.00 (25 °C, phosphate), pH 10.00(borate), Certipur®
Synonym(s):
Buffer solution pH 4.01 (25 °C, phthalate), pH 7.00 (25 °C, phosphate), pH 10.00(borate)
About This Item
Recommended Products
grade
certified reference material
Quality Level
Agency
traceable to NIST
traceable to PTB
product line
Certipur®
pH
10.00(borate)
4.01 (25 °C, phthalate)
7.00 (25 °C, phosphate)
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
Related Categories
General description
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to secondary reference standards from NIST and primary certified reference standards from PTB
- Ready-to-use solutions available in 30 mL sachets
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
- Packaging material subjected to comprehensive stability studies as per ISO Guide 35
- pH 4.00 and pH 6.99 buffer CRMs in the pack are accurate upto +/- 0.02 at 25 °C and the pH 10.01 buffer CRM is accurate upto +/-0.03 at 25 °C
Legal Information
recommended
Storage Class Code
10-13 - German Storage Class 10 to 13
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
Hi. I have a question: How can we use these buffers in similar pickets? Are they one-time use?
1 answer-
Product 1.99006, is Certipur® buffer solutions are single-use sachets. The instructions can be found in the link below:
https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/406/790/crm-buffer-solutions-ds8577en-ms.pdfIf additional information is needed. Please contact Technical Services directly.
Helpful?
-
-
This product is traceable to NIST, but does it comply with the SR EN 17034 standard?
1 answer-
This product is measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory. Please see the "Features and Benefits" section of the Product Detail Page for more information. While 'SR EN' 17034 is not specifically cited, full traceability and accreditation information is published in the product Certificate of Analysis.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service