Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.09438
Buffer solution
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 10.00 (20°C) Certipur®
Synonym(s):
Buffer solution pH 10.00 (20°C) (boric acid/potassium chloride/sodium hydroxide)
About This Item
Recommended Products
grade
certified reference material
Quality Level
product line
Certipur®
pH
10.0 (20 °C in H2O)
bp
110 °C/1013 hPa
mp
-6 °C
density
1.00 g/cm3 at 20 °C
application(s)
environmental
food and beverages
general analytical
industrial qc
pharmaceutical
format
mixture
storage temp.
15-25°C
General description
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Application
Features and Benefits
- Ready-to-use solution available in a sealed bottle and the innovative Titripac® packaging for positive environmental impact due to reduced waste generation
- Long-lasting with a shelf life of 3 years
- Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective audit
- Subjected to comprehensive stability studies in accordance with ISO Guide 35
- Measured and qualified in an ISO/IEC 17025 and ISO 17034 accredited laboratory
- Traceable to SRMs from NIST and primary certified reference standards from PTB
Legal Information
recommended
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
-
Hi, is the expanded uncertainty applicable to the other temperatures listed on the specifications datasheet ? Thanks,
1 answer-
The expanded uncertainty would be slightly different with changes in temperature.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service