Skip to Content
Merck
All Photos(1)

Key Documents

N-085

Supelco

Naphyrone-D5 hydrochloride solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

Naphyrone-D₅ Salt Solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H18D5NO · HCl
Molecular Weight:
322.88
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C(CC([2H])([2H])C([2H])([2H])[2H])N1CCCC1)C2=CC(C=CC=C3)=C3C=C2.[H]Cl

InChI

1S/C19H23NO.ClH/c1-2-7-18(20-12-5-6-13-20)19(21)17-11-10-15-8-3-4-9-16(15)14-17;/h3-4,8-11,14,18H,2,5-7,12-13H2,1H3;1H/i1D3,2D2;

InChI key

SIOVYKZRZLWQSY-LUIAAVAXSA-N

General description

Naphyrone is a synthetic cathinone found in numerous bath salt blends under street names such as NRG-1 and Energy-1. The designer drug emerged in late 2010 as a new legal high in the UK after the banning of mephedrone. This Certified Spiking Solution® is suitable for use as an internal standard in naphyrone testing methods for clinical toxicology, forensic analysis, and urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service