Skip to Content
Merck
All Photos(1)

Key Documents

M-150

Supelco

3,4-Methylenedioxypyrovalerone-d8 HCl (MDPV-d8 HCl) solution

100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

3,4-Methylenedioxypyrovalerone-d8 hydrochloride, MDPV-D8 HCl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16D8H13NO3 · HCl
CAS Number:
Molecular Weight:
319.85
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland); psicótropo (Spain); Decreto Lei 15/93: Tabela IIA (Portugal)

concentration

100 μg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C16H21NO3.ClH/c1-2-5-13(17-8-3-4-9-17)16(18)12-6-7-14-15(10-12)20-11-19-14;/h6-7,10,13H,2-5,8-9,11H2,1H3;1H/i3D2,4D2,8D2,9D2;

InChI key

PYQZNWFQAMFAMT-FQJBGNSISA-N

General description

An internal standard of 3,4-Methylenedioxypyrovalerone, a widely known bath salt and designer drug with cases of abuse reported throughout the US and Europe. Also known as MDPV, Mtv, or Super Coke, the illicit drug started appearing in the US in 2010 as a legal drug alternative sold under names including Ivory Wave, Tranquility, and Cloud 9.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Hofmann et al.
Molecular & cellular proteomics : MCP, 4(12), 1888-1897 (2005-08-23)
Identification of major histocompatibility complex (MHC)-associated peptides recognized by T-lymphocytes is a crucial prerequisite for the detection and manipulation of specific immune responses in cancer, viral infections, and autoimmune diseases. Unfortunately immunogenic peptides are less abundant species present in highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service