Key Documents
Z611182
Hirschmann® melting point determination tubes
capillary tubes, L × I.D. × O.D. 75 mm × 0.8 mm × 1 mm
About This Item
UNSPSC Code:
41112203
NACRES:
NB.79
manufacturer/tradename
Hirschmann
L × I.D. × O.D.
75 mm × 0.8 mm × 1 mm
Legal Information
Hirschmann is a registered trademark of Hirschmann Laborgeräte GmbH & Co.