Skip to Content
Merck
All Photos(1)

Key Documents

Z114952

Sigma-Aldrich

Capillary tube

tube capacity 1-5 μL

Synonym(s):

851321, Capillary Glass Tube, Capillary Tube, Glass Capillary Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.

material

glass
white

packaging

pack of 200 ea

manufacturer/tradename

WHEATON® 851321

tube capacity

1-5 μL

Looking for similar products? Visit Product Comparison Guide

General description

Borosilicate glass. For use with above positive displacement micropipettes.

Application

Capillarytube has been used to create a lung tissue phantom that mimics the alveolaranatomy.[1]

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Pacheco et al.
Journal of biomedical optics, 27(7) (2021-11-03)
Gas in scattering media absorption spectroscopy (GASMAS) enables noninvasive gas sensing in the body. It is developing as a tool for diagnosis and monitoring of respiratory conditions in neonates. Phantom models with relevant features to the clinical translation of GASMAS
Myung Gil Choi et al.
Scientific reports, 9(1), 3348-3348 (2019-03-06)
In this study, a novel resorufin thionocarbonate-based Hg2+-selective signaling probe (RT) for microfluidic paper-based analytical device (μPAD) applications is reported. The designed probe, RT, was readily synthesized by the one-step reaction of resorufin with phenyl thionochloroformate. The RT probe displayed

Questions

1–2 of 2 Questions  
  1. Can you please determine the outer diameter of the capillary tube Z114952?

    1 answer
    1. The capillary Tube (Product number Z114952) has an outer diameter of 1.25mm and an inner diameter of 0.5mm.

      Helpful?

  2. What is the inner diameter, outer diameter, and the length of the capillary tube?

    1 answer
    1. The dimensions for this product is as follows:
      Length (mm) x ID (mm) x OD (mm) 125mm x 0.5mm x 1.25mm

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service